Anti-PAR2 Mouse Monoclonal Antibody [clone: 7H40]
Catalog # USBIP3105-75E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:PAR2
- Clonality:Monoclonal
- Clone:7H40
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:lyophilized powder
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 25 ?g/ml
Type: Primary
Antigen: PAR2
Clonality: monoclonal
Clone: 7H40
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human