Anti-PACAP-Related Peptide Rabbit Polyclonal Antibody
Catalog # USBIP1790-35
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:PACAP Related Peptide
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Format:Serum
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:400-1:800
Type: Primary
Antigen: PACAP-Related Peptide
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat