Order Entry
Sweden
ContactUsLinkComponent
Anti-PA28, alpha Rabbit Polyclonal Antibody
Anti-PA28, alpha Rabbit Polyclonal Antibody
Catalog # USBIP1700-20
Supplier:  US Biological
Anti-PA28, alpha Rabbit Polyclonal Antibody
Catalog # USBIP1700-20
Supplier:  US Biological

Specifications

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    PA28, alpha
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • Reaktivitet:
    Hamster,
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, 50% glycerol and less than 0,02% sodium azide.
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    100 µl
  • Förp.:
    100 µl

Specifications

About this item

Type: Primary
Antigen: PA28, alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster, Human, Monkey, Mouse, Rat