Order Entry
Sweden
ContactUsLinkComponent
 
Anti-p40 phox Rabbit Polyclonal Antibody
  USBIP1001-20C
Anti-p40 phox Rabbit Polyclonal Antibody
  USBIP1001-20C

 

  • Antibody type:
    Primary
  • Antigen symbol:
    P40PHOX
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    0,1M Tris-glycine, pH 7,4, 0,15M sodium chloride, 0,05% sodium azide, before the addition of glycerol to 30%.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µg
  • Pk:
    200 µG

 

 

Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: p40 phox
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat