Order Entry
Sweden
ContactUsLinkComponent
 
Anti-CDKN1B Rabbit polyclonal antibody
  USBIP1001-05S
undefined
Anti-CDKN1B Rabbit polyclonal antibody
  USBIP1001-05S

 

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin-dependent kinase inhibitor 1B
  • Antigen symbol:
    CDKN1B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Modification:
    phospho-Thr187
  • Storage buffer:
    PBS, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: CDKN1B (cyclin-dependent kinase inhibitor 1B (p27, Kip1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human