Order Entry
Sweden
ContactUsLinkComponent
Anti-SLC5A5 Mouse Monoclonal Antibody [clone: 12F3]
Anti-SLC5A5 Mouse Monoclonal Antibody [clone: 12F3]
Catalog # USBIN2750-10A
Supplier:  US Biological
Anti-SLC5A5 Mouse Monoclonal Antibody [clone: 12F3]
Catalog # USBIN2750-10A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 5 (sodium/iodide cotransporter), member 5
  • Antigen symbol:
    Solute carrier family 5, member 5
  • Clonality:
    Monoclonal
  • Clone:
    12F3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    468 to 643
  • Storage buffer:
    PBS, pH 7,4, 0,09% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: SLC5A5 (solute carrier family 5 (sodium/iodide cotransporter), member 5)
Clonality: monoclonal
Clone: 12F3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat