Anti-Niemann-Pick Disease, Type C1 Gene-Like 1 Rabbit Polyclonal Antibody
Katalog # USBIN2566-32D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Niemann-Pick Disease, Type C1 Gene-Like 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 10mM sodium HEPES, pH 7,5, 150mM NaCl,0,1mg/ml BSA, 0,1% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: Niemann-Pick Disease, Type C1 Gene-Like 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human