Anti-Niemann-Pick Disease, Type C1 Gene-Like 1 Rabbit Polyclonal Antibody
Catalog # USBIN2566-32C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Niemann-Pick Disease, Type C1 Gene-Like 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Niemann-Pick Disease, Type C1 Gene-Like 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat