Order Entry
Sweden
ContactUsLinkComponent
Anti-Niemann-Pick Disease, Type C1 Gene-Like 1 Rabbit Polyclonal Antibody
Anti-Niemann-Pick Disease, Type C1 Gene-Like 1 Rabbit Polyclonal Antibody
Katalog # USBIN2566-32B
Supplier:  US Biological
undefined
Anti-Niemann-Pick Disease, Type C1 Gene-Like 1 Rabbit Polyclonal Antibody
Katalog # USBIN2566-32B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Niemann-Pick Disease, Type C1 Gene-Like 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Pig,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    Tris-citrate/phosphate, pH 7,5, 0,1% sodium azide
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:500; Immunofluorescence: 1:100-1:250; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: Niemann-Pick Disease, Type C1 Gene-Like 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Pig, Rat