Anti-NgCAM Related Neuronal Cell Adhesion Molecule Mouse Monoclonal Antibody [clone: 7H74]
Catalog # USBIN2412
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:NgCAM Related Neuronal Cell Adhesion Molecule
- Clonality:Monoclonal
- Clone:7H74
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:30 to 600
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: NgCAM Related Neuronal Cell Adhesion Molecule
Clonality: monoclonal
Clone: 7H74
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human