Anti-Neurokinin B Receptor Rabbit Polyclonal Antibody
USBIN2171-60A
- Antibody type:Primary
- Antigen name:Neurokinin B Receptor
- Antigen symbol:NKBR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunochemistry: 5 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: Neurokinin B Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse