Order Entry
Sweden
ContactUsLinkComponent
 
Anti-MEF2C Rabbit Polyclonal Antibody (FITC (Fluorescein))
  USBIM9769-97E-FITC
undefined
Anti-MEF2C Rabbit Polyclonal Antibody (FITC (Fluorescein))
  USBIM9769-97E-FITC

 

  • Antibody type:
    Primary
  • Antigen name:
    Myocyte enhancer factor 2C
  • Antigen symbol:
    MEF2C
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human