Anti-MHC Class 1 Chain-related Gene A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBIM3886-75A-HRP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 1 Chain-related Gene A
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:75 to 105
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:50; Immunofluorescence: 1:10-1:50
Type: Primary
Antigen: MHC Class 1 Chain-related Gene A
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human