Anti-MGMT Chicken Polyclonal Antibody
Catalog # USBIM3884-92A
Leverantör: US Biological
Specifikationer
- Antibody type:Primary
- Antigen name:O-6-methylguanine-DNA methyltransferase
- Antigen symbol:MGMT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 207
- Storage buffer:PBS, pH 7,2, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifikationer
Om denna produkt
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: MGMT (O-6-methylguanine-DNA methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human