Anti-Melatonin Rabbit Polyclonal Antibody
Catalog # USBIM2871-04E
Leverantör: US Biological
Specifikationer
- Antibody type:Primary
- Antigen name:Melatonin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by protein carrier (BSA) affinity column, and ammonium sulphate precipitation
- Size:100 µg
- Pk:100 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: Melatonin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: