Anti-MAS-Related G-Protein Coupled Receptor, Member C Rabbit Polyclonal Antibody
USBIM2412-34A
- Antibody type:Primary
- Antigen name:MAS-Related G-Protein Coupled Receptor, Member C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:14 to 28
- Storage buffer:PBS, pH 7,4, 5% BSA, 0,05% thimerosal, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 0.75 ?g/ml; Immunochemistry: 0.5-1 ?g/ml
Type: Primary
Antigen: MAS-Related G-Protein Coupled Receptor, Member C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat