Order Entry
Sweden
ContactUsLinkComponent
Anti-Mas-related G-protein Coupled Receptor Member X1 Rabbit Polyclonal Antibody
Anti-Mas-related G-protein Coupled Receptor Member X1 Rabbit Polyclonal Antibody
Catalog # USBIM2412-34
Supplier:  US Biological
Anti-Mas-related G-protein Coupled Receptor Member X1 Rabbit Polyclonal Antibody
Catalog # USBIM2412-34
Supplier:  US Biological

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Mas-related G-protein Coupled Receptor Member X1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Rat
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Aminosyretal:
    14 to 28
  • Storage buffer:
    PBS, 5% BSA, 0,05% thimerosal, 0,05% sodium azide.
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    ~1 mg/ml (after reconstitution)
  • Shipping temperature:
    Ice
  • Rening:
    purified by affinity chromatography
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 0.1-0.5 ?g/ml; Immunochemistry: 0.5-1.0 ?g/ml

Type: Primary
Antigen: Mas-related G-protein Coupled Receptor Member X1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat