Anti-MAP Kinase 1 Mouse Monoclonal Antibody
Catalog # USBIM2351-80S
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MAP Kinase 1
- Antigen symbol:MAPK1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:HEPES, 0,15M sodium chloride, 0,01% BSA, 0,03% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:50
Type: Primary
Antigen: MAP Kinase 1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human