Anti-Lysine, Acetylated Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIL9058-05A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Acetyl-Lysine
- Antigen symbol:AcK
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH7, 50% glycerol.
- Storage temperature:Cool
- Concentration:~0,25 mg/ml
- Shipping temperature:Ice
- Tested applications:Radio Immunoassay
- Purification:purified by immunoaffinity chromatography
- Size:400 µl
- Pk:400 µl
Specifications
About this item
Recommended dilution: Western Blot:1:250
Type: Primary
Antigen: Lysine, Acetylated
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: