Anti-LOX-1, Soluble form Mouse Monoclonal Antibody [clone: 9L452]
USBIL2623-01E
- Antibody type:Primary
- Antigen name:LOX-1, Soluble form
- Antigen symbol:LOX-1
- Clonality:Monoclonal
- Clone:9L452
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: LOX-1, Soluble form
Clonality: monoclonal
Clone: 9L452
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human