Anti-Leucine Rich Repeat Transmembrane Protein 2 Sheep Polyclonal Antibody
Catalog # USBIL2020-76A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Leucine Rich Repeat Transmembrane Protein 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:34 to 422
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: Leucine Rich Repeat Transmembrane Protein 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse