Anti-Kv3.1b Mouse Monoclonal Antibody [clone: 10F749]
Catalog # USBIK9010-01
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:KV3.1B
- Clonality:Monoclonal
- Clone:10F749
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:437 to 585
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-10 ?g/ml; Immunochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: Kv3.1b
Clonality: monoclonal
Clone: 10F749
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat