Anti-KREMEN1 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIK5000-01-BIOT
Supplier: US Biological
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:kringle containing transmembrane Protein 1
- Antigensymbol:KREMEN1
- Klonalitet:Polyclonal
- Konjugation:Biotin
- ELISA:Yes
- Värd:Rabbit
- Isotyp:IgG
- Reaktivitet:Human
- Western blot:Yes
- Epitop:N-terminal
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:200 µl
- Förp.:200 µl
Specifications
About this item
Type: Primary
Antigen: KREMEN1 (kringle containing transmembrane protein 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human