Order Entry
Sweden
ContactUsLinkComponent
 
Anti-Kinase-interacting Protein 2 Rabbit Polyclonal Antibody
  USBIK1775-75N
undefined
Anti-Kinase-interacting Protein 2 Rabbit Polyclonal Antibody
  USBIK1775-75N

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Kinase-interacting Protein 2
  • Antigensymbol:
    AKIP2
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    N-terminal
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,09% sodium azide.
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Rening:
    purified by Protein G chromatography
  • Storlek:
    200 µl
  • Förp.:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: Kinase-interacting Protein 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human