Anti-KLH Mouse Monoclonal Antibody
Catalog # USBIK0300-03D
Supplier: US Biological
Specifications
- Antibody type:Tag
- Antigen name:Keyhole Limpet Hemocyanin
- Antigen symbol:KLH
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-2?g/ml
Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: