Anti-Interferon Stimulating Gene 15 Rabbit Monoclonal Antibody [clone: 7H28]
Catalog # USBII7662-94A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interferon Stimulating Gene 15
- Clonality:Monoclonal
- Clone:7H28
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Horse,Human
- Western blot:Yes
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:25
Type: Primary
Antigen: Interferon Stimulating Gene 15
Clonality: monoclonal
Clone: 7H28
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Horse, Human