Anti-Interferon Stimulating Gene 15 Rabbit Polyclonal Antibody
Catalog # USBII7662-94
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interferon Stimulating Gene 15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~5 mg/ml
- Shipping temperature:Ice
- Purification:purified by delipidation, salt fractionation and ion exchange chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Recommended dilution: ELISA: 1:2000-1:10000; Western Blot: 1:200-1:1000
Type: Primary
Antigen: Interferon Stimulating Gene 15
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human