Anti-Interferon gamma Rabbit Polyclonal Antibody
USBII7661-68M
- Antibody type:Primary
- Antigen name:Interferon gamma
- Antigen symbol:IFN gamma
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Chicken
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: Interferon gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken