Anti-Interferon gamma Mouse Monoclonal Antibody [clone: GF1]
USBII7661-66S
- Antibody type:Primary
- Antigen name:Interferon gamma
- Antigen symbol:IFN gamma
- Clonality:Monoclonal
- Clone:GF1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, containing 0,1% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Interferon gamma
Clonality: monoclonal
Clone: GF1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human