Order Entry
Sweden
ContactUsLinkComponent
Anti-Inositol 1,4,5-trisphosphate 3-kinase A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anti-Inositol 1,4,5-trisphosphate 3-kinase A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Katalog # USBII7655-65C-AP
Supplier:  US Biological
undefined
Anti-Inositol 1,4,5-trisphosphate 3-kinase A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Katalog # USBII7655-65C-AP
Supplier:  US Biological

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Inositol 1,4,5-trisphosphate 3-kinase A
  • Antigensymbol:
    ITP3K
  • Klonalitet:
    Polyclonal
  • Konjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Rening:
    purified by Protein G chromatography
  • Storlek:
    200 µl
  • Förp.:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: Inositol 1,4,5-trisphosphate 3-kinase A
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse