Order Entry
Sweden
ContactUsLinkComponent
Anti-ICOSLG Mouse Monoclonal Antibody [clone: 8H103]
Anti-ICOSLG Mouse Monoclonal Antibody [clone: 8H103]
Catalog # USBII0650-01H
Supplier:  US Biological
Anti-ICOSLG Mouse Monoclonal Antibody [clone: 8H103]
Catalog # USBII0650-01H
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    inducible T-cell co-stimulator ligand
  • Antigen symbol:
    ICOSLG
  • Clonality:
    Monoclonal
  • Clone:
    8H103
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Neutralisation:
    Yes
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, 5% trehalose.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Type: Primary
Antigen: ICOSLG (inducible T-cell co-stimulator ligand)
Clonality: monoclonal
Clone: 8H103
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human