Anti-Histone H4, acetylated Rabbit polyclonal antibody
USBIH5110-15F1
- Antibody type:Primary
- Antigen name:Histone H4
- Antigen symbol:H4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- ChIP:Yes
- Format:Serum
- Form:liquid
- Amino acid number:3 to 13
- Modification:acetyl-Lys8
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Chromatin, Dot Blot
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000; ChImmunoprecipitation: 0.1-0.5?l
Type: Primary
Antigen: Histone H4, acetylated
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human