Anti-G Protein Coupled Receptor 8 Rabbit polyclonal antibody
Catalog # USBIG8600-12B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:G Protein Coupled Receptor 8
- Antigen symbol:GPR8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:319 to 333
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: G Protein Coupled Receptor 8
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human