Anti-GITRL Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 11n77]
USBIG2035-15C
- Antibody type:Primary
- Antigen symbol:GITRL
- Clonality:Monoclonal
- Clone:11n77
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:saline, 0,5% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,025 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Do not freeze!
Type: Primary
Antigen: GITRL
Clonality: monoclonal
Clone: 11n77
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human