Anti-FMN2 Rabbit polyclonal antibody
Catalog # USBIF5385-01A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Formin 2
- Antigen symbol:FMN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Format:Serum
- Form:liquid
- Amino acid number:168 to 186
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Immunochemistry: 1:100
Type: Primary
Antigen: FMN2 (formin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse