Anti-Feline Immunodeficiency Virus, envelope, gp95 Mouse monoclonal antibody [clone: 9E39]
Catalog # USBIF0030-18A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Feline Immunodeficiency Virus, envelope, gp95
- Clonality:Monoclonal
- Clone:9E39
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,05% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Type: Primary
Antigen: Feline Immunodeficiency Virus, envelope, gp95
Clonality: monoclonal
Clone: 9E+39
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: