Anti-EN1 Mouse monoclonal antibody [clone: 1F5]
USBIE2270-02A
- Antibody type:Primary
- Antigen name:engrailed homeobox 1
- Antigen symbol:EN1
- Clonality:Monoclonal
- Clone:1F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:266 to 392
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: EN1 (engrailed homeobox 1)
Clonality: monoclonal
Clone: 1F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human