Anti-Dynamin Mouse Monoclonal Antibody [clone: 4E67]
USBID9900-01-100
- Antibody type:Primary
- Antigen name:Dynamin
- Antigen symbol:DNM
- Clonality:Monoclonal
- Clone:4E67
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Chicken,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Dynamin
Clonality: monoclonal
Clone: 4E+67
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chicken, Human, Mouse, Rat