Order Entry
Sweden
ContactUsLinkComponent
Anti-Corticotropin-Releasing Factor Receptor 1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H27]
Anti-Corticotropin-Releasing Factor Receptor 1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H27]
  USBIC7903-89B
Anti-Corticotropin-Releasing Factor Receptor 1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H27]
  USBIC7903-89B

 

  • Antibody type:
    Primary
  • Antigen name:
    Corticotropin releasing factor receptor 1
  • Antigen symbol:
    CRF1
  • Clonality:
    Monoclonal
  • Clone:
    7H27
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 Tests
  • Pk:
    100 Tests

 

 

Do not freeze!

Type: Primary
Antigen: Corticotropin-Releasing Factor Receptor 1
Clonality: Monoclonal
Clone: 7H27
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human