Anti-Ceramide Mouse monoclonal antibody [clone: 6D311]
USBIC2777-95
- Antibody type:Primary
- Antigen name:Ceramide
- Clonality:Monoclonal
- Clone:6D311
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Pig,Monkey,Sheep,Goat,Dog,Rat
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by gel filtration on sephacryl S-300
- Size:50 Tests
- Pk:50 Tests
Recommended dilution: ELISA: 1:10; Flow Cytometry: 20?l/test; Immunochemistry: 1:10
Type: Primary
Antigen: Ceramide
Clonality: Monoclonal
Clone: 6D311
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Dog, Goat, Monkey, Pig, Rat, Sheep