Anti-CDC42BPA Mouse Monoclonal Antibody [clone: 10B2176]
Catalog # USBIC2562-83
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CDC42 binding protein kinase alpha (DMPK-like)
- Antigen symbol:CDC42BPA
- Clonality:Monoclonal
- Clone:10B2176
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:551 to 650
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:500-1:1000; ELISA: 1 ?g/ml-3ng/ml
Type: Primary
Antigen: CDC42BPA (CDC42 binding protein kinase alpha (DMPK-like))
Clonality: Monoclonal
Clone: 10B2176
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human