Order Entry
Sweden
ContactUsLinkComponent
Anti-IFNGR1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: BB1E2]
Anti-IFNGR1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: BB1E2]
Katalog # USBIC2453-01F
Leverantör:  US Biological
Anti-IFNGR1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: BB1E2]
Katalog # USBIC2453-01F
Leverantör:  US Biological

Specifikationer

  • Antibody type:
    Primary
  • Antigen name:
    Interferon gamma receptor 1
  • Antigen symbol:
    IFNGR1
  • Clonality:
    Monoclonal
  • Clone:
    BB1E2
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2 0,09% sodium azide
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifikationer

Om denna produkt

Recommended dilution: Flow Cytometry: 1-10 ?g/ml; FLISA: 1 ?g/ml; Western Blot: 1 ?g/ml; Immunochemistry: 1-10 ?g/ml

Type: Primary
Antigen: IFNGR1 (interferon gamma receptor 1)
Clonality: Monoclonal
Clone: BB1E2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human