Anti-FAS Mouse Monoclonal Antibody [clone: B-R18]
Catalog # USBIC2442-21F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fas cell surface death receptor
- Antigen symbol:FAS
- Clonality:Monoclonal
- Clone:B-R18
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: Monoclonal
Clone: B-R18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: