Anti-CD53 Rat Monoclonal Antibody [clone: 3H1542]
USBIC2408-11B
- Antibody type:Primary
- Antigen name:CD53 molecule
- Antigen symbol:CD53
- Clonality:Monoclonal
- Clone:3H1542
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgM
- Reactivity:Mouse
- Format:Tissue culture supernatant
- Form:liquid
- Storage temperature:Cool
- Concentration:~15 mg/ml
- Shipping temperature:Ice
- Size:2 ml
- Pk:2 mL
Type: Primary
Antigen: CD53 (CD53 molecule)
Clonality: Monoclonal
Clone: 3H1542
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse