Anti-CD45RC Rat Monoclonal Antibody FLMA [clone: 5K258]
Catalog # USBIC2400-61D
Leverantör: US Biological
Specifikationer
- Antibody type:Primary
- Antigen symbol:CD45RC
- Clonality:Monoclonal
- Clone:5K258
- Conjugation:FLMA (Fluorescein-5-Maleimide)
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgM kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: CD45RC
Clonality: Monoclonal
Clone: 5K258
Conjugation: FLMA
Epitope:
Host: Rat
Isotype: IgM Kappa
Reactivity: Mouse