Anti-CD14, MONOCYTE, MACROPHAGE MARKER Mouse Monoclonal Antibody [clone: 9G141]
Catalog # USBIC2265-51A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD14 Monocytes
- Antigen symbol:CD14
- Clonality:Monoclonal
- Clone:9G141
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig,Human
- Form:liquid
- Storage buffer:PBS, 0,1% BSA.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:50
Type: Primary
Antigen: CD14, MONOCYTE, MACROPHAGE MARKER
Clonality: Monoclonal
Clone: 9G141
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig