Order Entry
Sweden
ContactUsLinkComponent
Anti-SCAF11 Rabbit Polyclonal Antibody
Anti-SCAF11 Rabbit Polyclonal Antibody
Catalog # USBIC2088-56E
Supplier:  US Biological
Anti-SCAF11 Rabbit Polyclonal Antibody
Catalog # USBIC2088-56E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SR-related CTD-associated factor 11
  • Antigen symbol:
    SCAF11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    lyophilized powder
  • Amino acid number:
    286 to 298
  • Storage buffer:
    PBS, 0,1% BSA.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml (after reconstitution)
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1-10 ?g/ml; ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: SCAF11 (SR-related CTD-associated factor 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse