Anti-Caspase 3, active Rabbit Polyclonal Antibody
USBIC2087-22L
- Antibody type:Primary
- Antigen name:Caspase 3, active
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,5% BSA, 0,01% thimerosal, 30% glycerol.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunochemistry: 1:10-1:20; Western Blot: 1:50-1:500
Type: Primary
Antigen: Caspase 3, active
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat