Anti-Carnitine O-palmitoyltransferase 1, Brain Isoform Rabbit polyclonal antibody
USBIC1385-29X
- Antibody type:Primary
- Antigen name:Carnitine O-palmitoyltransferase 1, Brain Isoform
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Form:lyophilized powder
- Amino acid number:769 to 798
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Carnitine O-palmitoyltransferase 1, Brain Isoform
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse