Order Entry
Sweden
ContactUsLinkComponent
 
Anti-CPA1 Rabbit Polyclonal Antibody
  USBIC1200-01D
Anti-CPA1 Rabbit Polyclonal Antibody
  USBIC1200-01D

 

  • Antibody type:
    Primary
  • Antigen name:
    carboxypeptidase A1 (pancreatic)
  • Antigen symbol:
    CPA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Bovine
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,01% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    50 µl
  • Pk:
    50 µl

 

 

Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CPA1 (carboxypeptidase A1 (pancreatic))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine